Please login to your online account to display your discounted pricing

1-Bromo-5-phenylpentane 98%, ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC384740010

 View more versions of this product

Catalog No. AC384740010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Formula Weight 227.14
Physical Form Liquid
Quantity 1g
Color Clear colorless
Density 1.23g/mL
CAS Max % 100.0
Linear Formula C6H5(CH2)5Br
Molecular Formula C11H15Br
CAS 14469-83-1
CAS Min % 97.5
MDL Number MFCD01075177
Packaging Glass bottle
Infrared Spectrum Authentic
Refractive Index 1.5320 to 1.5340 (20°C, 589nm)
Synonym (5-Bromopentyl)benzene
Chemical Name or Material 1-Bromo-5-phenylpentane
Assay Percent Range 98%
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.

Specifications

Chemical Name or Material 1-Bromo-5-phenylpentane
CAS 14469-83-1
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C11H15Br
Linear Formula C6H5(CH2)5Br
Formula Weight 227.14
MDL Number MFCD01075177
Synonym (5-Bromopentyl)benzene
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.

SDS & Safety Documents