Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Bromo-5-methylhexane, 97%

Chemical Identifiers

Chemical Name or Material 1-Bromo-5-methylhexane
CAS 35354-37-1
Molecular Formula C7H15Br
Formula Weight 179.11
MDL Number MFCD00041674
UN Number UN1993
Beilstein 1731802
Synonym 5-Methylhexyl bromide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0948603

  SDS
alfa aesar™
L0948603
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL0948606

  SDS
alfa aesar™
L0948606
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 179.11
Molecular Formula C7H15Br
Chemical Name or Material 1-Bromo-5-methylhexane
CAS 35354-37-1
Synonym 5-Methylhexyl bromide
Health Hazard 3 GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Boiling Point 162° to 163°C
Beilstein 1731802
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Assay 97%
DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
UN Number UN1993
Quantity 1g
Flash Point 57°C (134°F)
MDL Number MFCD00041674

Applications
1-Bromo-5-methylhexane reacts with 3-fluoro-dodec-1-yne to produce 9-fluoro-2-methyl-octadec-7-yne. It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff.

Solubility
Not miscible with water.

Notes
Incompatible with strong oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.