Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Bromo-5-fluoropentane, 99%

Chemical Identifiers

Chemical Name or Material 1-Bromo-5-fluoropentane
CAS 407-97-6
Linear Formula F(CH2)5Br
Formula Weight 169.04
MDL Number MFCD01709395
Beilstein 1697258
Synonym 5-Fluoroamyl bromide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6480603

  SDS
alfa aesar™
H6480603
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6480606

  SDS
alfa aesar™
H6480606
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH64806MD

  SDS
alfa aesar™
H64806MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 169.04
Chemical Name or Material 1-Bromo-5-fluoropentane
Refractive Index 1.4406
Density 1.360g/mL
Color Colorless
Physical Form Liquid
Assay 99%
Boiling Point 162°C
Quantity 1g
Linear Formula F(CH2)5Br
MDL Number MFCD01709395
CAS 407-97-6
Beilstein 1697258
Synonym 5-Fluoroamyl bromide
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
RTECS Number RZ9810000

Applications
1-Bromo-5-fluoropentane was used to prepare 18F-labeled (anilido)piperidine derivatives as μ-opioid receptor imaging agents.

Solubility
Soluble in methanol (in methanol). Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.