Please login to your online account to display your discounted pricing

1-Bromo-4-tert-butylbenzene, 97%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC225190050

 View more versions of this product

Catalog No. AC225190050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥96.0%
Physical Form Liquid
Formula Weight 213.12
Quantity 5g
Color Clear colorless to slightly yellow
EINECS Number 223-599-2
TSCA TSCA
Density 1.22g/mL
Beilstein 05, 416
CAS Min % 96.0
CAS Max % 100.0
Molecular Formula C10H13Br
Packaging Glass bottle
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Refractive Index 1.5315 to 1.5345 (20°C, 589nm)
Name Note 97%
Assay Percent Range 97%
Linear Formula (CH3)3CC6H4Br
MDL Number MFCD00000108
CAS 3972-65-4
Chemical Name or Material 1-Bromo-4-tert-butylbenzene
Infrared Spectrum Authentic
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

Specifications

Chemical Name or Material 1-Bromo-4-tert-butylbenzene
Name Note 97%
CAS 3972-65-4
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C10H13Br
Linear Formula (CH3)3CC6H4Br
Formula Weight 213.12
MDL Number MFCD00000108
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

SDS & Safety Documents