Please login to your online account to display your discounted pricing

1-Bromo-4-propylbenzene, 99%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 1-Bromo-4-propylbenzene
CAS 588-93-2
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C9H11Br
Linear Formula CH3CHCHC6H4Br
Formula Weight 199.1
MDL Number MFCD00012456
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC308240050

  SDS
thermo fisher scientific
308240050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥98.5%
Physical Form Liquid
Formula Weight 199.1
Quantity 5g
Color Clear colorless to slightly yellow
CAS Max % 100.0
Chemical Name or Material 1-Bromo-4-propylbenzene
Assay Percent Range 99%
Molecular Formula C9H11Br
MDL Number MFCD00012456
CAS 588-93-2
CAS Min % 98.5
Infrared Spectrum Authentic
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Packaging Glass bottle
Refractive Index 1.5350 to 1.5370 (20°C, 589nm)
Linear Formula CH3CHCHC6H4Br
Health Hazard 2 GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your 9-digit account number, or become a registered user on fishersci.com by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service at 1-800-766-7000.