missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
1-Bromo-4-phenylbutane, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals B2523706
Description
1-Bromo-4-phenylbutane is used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1-Bromo-4-phenylbutane is used as pharmaceutical intermediate.
Solubility
Slightly soluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications
1-Bromo-4-phenylbutane | |
1.214 | |
C10H13Br | |
5 g | |
Slightly soluble in water. | |
BrCCCCC1=CC=CC=C1 | |
213.12 | |
213.12 |
13633-25-5 | |
131°C to 133°C (12 mmHg) | |
MFCD00154988 | |
1-bromo-4-phenylbutane, 4-bromobutyl benzene, 4-phenylbutyl bromide, 4-phenyl 1-butyl bromide, 4-phenyl-1-butyl bromide, 4-phenyl-1-bromobutane, 1-bromo-4-phenyl-butane, benzene, 4-bromobutyl, benzene, bromobutyl, 4-phenyl-1-bromo butane | |
XPBQQAHIVODAIC-UHFFFAOYSA-N | |
4-bromobutylbenzene | |
259668 | |
98% |
Chemical Identifiers
13633-25-5 | |
213.12 | |
XPBQQAHIVODAIC-UHFFFAOYSA-N | |
259668 |
C10H13Br | |
MFCD00154988 | |
1-bromo-4-phenylbutane, 4-bromobutyl benzene, 4-phenylbutyl bromide, 4-phenyl 1-butyl bromide, 4-phenyl-1-butyl bromide, 4-phenyl-1-bromobutane, 1-bromo-4-phenyl-butane, benzene, 4-bromobutyl, benzene, bromobutyl, 4-phenyl-1-bromo butane | |
BrCCCCC1=CC=CC=C1 |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only