Please login to your online account to display your discounted pricing

1-Bromo-4-pentylbenzene, 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 1-Bromo-4-pentylbenzene
CAS 51554-95-1
Assay Percent Range 97%
Molecular Formula C11H15Br
Linear Formula BrC6H4(CH2)4CH3
MDL Number MFCD00061113
Synonym 4-Pentylbromobenzene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC399460050

  SDS
thermo fisher scientific
AC399460050
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC399460010

  SDS
thermo fisher scientific
AC399460010
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Quantity 5g
Density 1.272g/cm3
Synonym 4-Pentylbromobenzene
Linear Formula BrC6H4(CH2)4CH3
Chemical Name or Material 1-Bromo-4-pentylbenzene
Assay Percent Range 97%
CAS 51554-95-1
Molecular Formula C11H15Br
MDL Number MFCD00061113