Please login to your online account to display your discounted pricing

1-Bromo-4-pentylbenzene, 97%, ACROS Organics™


Quantity 1g

Manufacturer: thermo fisher scientific  AC399460010

 View more versions of this product

Catalog No. AC399460010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Quantity 1g
Chemical Name or Material 1-Bromo-4-pentylbenzene
Assay Percent Range 97%
Linear Formula BrC6H4(CH2)4CH3
CAS 51554-95-1
Molecular Formula C11H15Br
MDL Number MFCD00061113
Synonym 4-Pentylbromobenzene

Specifications

Chemical Name or Material 1-Bromo-4-pentylbenzene
CAS 51554-95-1
Assay Percent Range 97%
Molecular Formula C11H15Br
Linear Formula BrC6H4(CH2)4CH3
MDL Number MFCD00061113
Synonym 4-Pentylbromobenzene