Learn More
1-Bromo-4-nitrobenzene, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1770536
Description
It can be employed as a syntheses material intermediate. It is also used as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt can be employed as a syntheses material intermediate. It is also used as a pharmaceutical intermediate.
Solubility
Insoluble in water. Almost transparent in hot ethyl alcohol. Soluble in ether
Notes
Store away from oxidizing agents and bases. Keep the container tightly closed in a cool, dry place.
Specifications
1-Bromo-4-nitrobenzene | |
124°C to 127°C | |
254°C to 256°C | |
MFCD00007280 | |
UN3459 | |
4-bromonitrobenzene, 4-nitrobromobenzene, p-bromonitrobenzene, benzene, 1-bromo-4-nitro, p-nitrobromobenzene, 4-nitrophenyl bromide, 4-bromo-1-nitrobenzene, p-nitrophenyl bromide, 1-bromo-4-nitro-benzene, ccris 3115 | |
ZDFBKZUDCQQKAC-UHFFFAOYSA-N | |
1-bromo-4-nitrobenzene | |
11466 | |
98% |
586-78-7 | |
1.948 | |
C6H4BrNO2 | |
500 g | |
636964 | |
Insoluble in water. Almost transparent in hot ethyl alcohol. Soluble in ether | |
[O-][N+](=O)C1=CC=C(Br)C=C1 | |
202.01 | |
202.01 |
Chemical Identifiers
586-78-7 | |
202.01 | |
ZDFBKZUDCQQKAC-UHFFFAOYSA-N | |
11466 | |
[O-][N+](=O)C1=CC=C(Br)C=C1 |
C6H4BrNO2 | |
MFCD00007280 | |
4-bromonitrobenzene, 4-nitrobromobenzene, p-bromonitrobenzene, benzene, 1-bromo-4-nitro, p-nitrobromobenzene, 4-nitrophenyl bromide, 4-bromo-1-nitrobenzene, p-nitrophenyl bromide, 1-bromo-4-nitro-benzene, ccris 3115 | |
1-bromo-4-nitrobenzene |
Safety and Handling
GHS H Statement
H331-H373-H302-H312-H315-H319-H335
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P260-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P314-P330-P332+P313-P362-P501c
H302+H312+H332-H315-H319-H335-H373
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITROBROMOBENZENES, SOLID
EINECSNumber : 209-583-8
RTECSNumber : CY9040550
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only