Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Bromo-4-(n-octyloxy)benzene, 98%

$48.97 - $575.58

Chemical Identifiers

Chemical Name or Material 1-Bromo-4-(n-octyloxy)benzene
CAS 96693-05-9
Molecular Formula C14H21BrO
Formula Weight 285.23
MDL Number MFCD00043445
Synonym 4-Bromophenyl n-octyl ether; 4-(n-Octyloxy)bromobenzene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1333403

  SDS
alfa aesar™
A1333403
1g Each for $48.97

AAA1333406

  SDS
alfa aesar™
A1333406
5g Each for $147.51

AAA1333414

  SDS
alfa aesar™
A1333414
25g Each for $575.58
Description & Specifications

Specifications

Formula Weight 285.23
Quantity 1g
Molecular Formula C14H21BrO
Synonym 4-Bromophenyl n-octyl ether; 4-(n-Octyloxy)bromobenzene
Boiling Point 135° to 137°C (0.5mmHg)
CAS 96693-05-9
Assay 98%
Chemical Name or Material 1-Bromo-4-(n-octyloxy)benzene
MDL Number MFCD00043445