Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Bromo-4-n-heptyloxybenzene, 97%

$57.57 - $203.42

Chemical Identifiers

Chemical Name or Material 1-Bromo-4-n-heptyloxybenzene
CAS 123732-04-7
Molecular Formula C13H19BrO
Formula Weight 271.2
MDL Number MFCD00173752
Synonym 4-n-Heptyloxybromobenzene; 1-(4-Bromophenoxy)heptane
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2484403

  SDS
alfa aesar™
B2484403
1g Each for $57.57

AAB2484406

  SDS
alfa aesar™
B2484406
5g Each for $203.42
Description & Specifications

Specifications

Formula Weight 271.2
Chemical Name or Material 1-Bromo-4-n-heptyloxybenzene
Assay 97%
Molecular Formula C13H19BrO
MDL Number MFCD00173752
CAS 123732-04-7
Synonym 4-n-Heptyloxybromobenzene; 1-(4-Bromophenoxy)heptane
Quantity 1g