missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Bromo-4-n-heptylbenzene, 97%

Manufacturer:  Alfa Aesar™ A1223506

 View more versions of this product

Catalog No. AAA1223506



Specifications

Specifications

1-Bromo-4-n-heptylbenzene
76287-49-5
MFCD00061115
YHKMTIJHJWYYAG-UHFFFAOYSA-N
1-bromo-4-heptylbenzene
2775100
97%
5g
C13H19Br
1-bromo-4-n-heptylbenzene, 1-4-bromophenyl heptane, 4-heptylbromobenzene, benzene, 1-bromo-4-heptyl, 4-bromo-1-heptylbenzene, 1-bromo-4-heptyl-benzene, acmc-209p2u, 1-bromanyl-4-heptyl-benzene, ksc912e9b
CCCCCCCC1=CC=C(C=C1)Br
255.199
255.2

Chemical Identifiers

76287-49-5
255.199
YHKMTIJHJWYYAG-UHFFFAOYSA-N
2775100
CCCCCCCC1=CC=C(C=C1)Br
C13H19Br
MFCD00061115
1-bromo-4-n-heptylbenzene, 1-4-bromophenyl heptane, 4-heptylbromobenzene, benzene, 1-bromo-4-heptyl, 4-bromo-1-heptylbenzene, 1-bromo-4-heptyl-benzene, acmc-209p2u, 1-bromanyl-4-heptyl-benzene, ksc912e9b
1-bromo-4-heptylbenzene
Safety and Handling

Safety and Handling

TSCA : TSCA

SDS Documents