missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Bromo-4-n-decylbenzene, 95%

$45.49 - $820.41

Chemical Identifiers

CAS 106418-67-1
Molecular Formula C16H25Br
Molecular Weight (g/mol) 297.28
MDL Number MFCD00061112
InChI Key SWTGVSCSSDYINB-UHFFFAOYSA-N
Synonym 1-bromo-4-n-decylbenzene, benzene, 1-bromo-4-decyl, 4-decylbromobenzene, acmc-2098jm, ksc909c2n
PubChem CID 13757384
IUPAC Name 1-bromo-4-decylbenzene
SMILES CCCCCCCCCCC1=CC=C(C=C1)Br
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6546503 View Documents Alfa Aesar™
H6546503
1g Each for $45.49
 
AAH6546506 View Documents Alfa Aesar™
H6546506
5g Each for $197.39
 
AAH6546514 View Documents Alfa Aesar™
H6546514
25g Each for $820.41
 
Specifications

Chemical Identifiers

106418-67-1
297.28
SWTGVSCSSDYINB-UHFFFAOYSA-N
13757384
CCCCCCCCCCC1=CC=C(C=C1)Br
C16H25Br
MFCD00061112
1-bromo-4-n-decylbenzene, benzene, 1-bromo-4-decyl, 4-decylbromobenzene, acmc-2098jm, ksc909c2n
1-bromo-4-decylbenzene

Specifications

1.102g/mL
Colorless to Yellow
106418-67-1
MFCD00061112
1-bromo-4-n-decylbenzene, benzene, 1-bromo-4-decyl, 4-decylbromobenzene, acmc-2098jm, ksc909c2n
CCCCCCCCCCC1=CC=C(C=C1)Br
297.28
297.28
95%
1.5110
1g
C16H25Br
4743413
SWTGVSCSSDYINB-UHFFFAOYSA-N
1-bromo-4-decylbenzene
13757384
Liquid
1-Bromo-4-n-decylbenzene