Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Bromo-4-isopropylbenzene, 97%

Chemical Identifiers

Chemical Name or Material 1-Bromo-4-isopropylbenzene
CAS 586-61-8
Molecular Formula C9H11Br
Formula Weight 199.1
MDL Number MFCD00039159
UN Number UN3082
Beilstein 1858096
Synonym 4-Bromocumene; 4-Isopropylbromobenzene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1337006

  SDS
alfa aesar™
A1337006
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1337009

  SDS
alfa aesar™
A1337009
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1337018

  SDS
alfa aesar™
A1337018
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 199.1
Boiling Point 219°C
Molecular Formula C9H11Br
TSCA TSCA
Chemical Name or Material 1-Bromo-4-isopropylbenzene
Melting Point −12°C
DOT Information Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCE, LIQUID, N.O.S.
Quantity 5g
Synonym 4-Bromocumene; 4-Isopropylbromobenzene
Flash Point >100°C (212°F)
Assay 97%
MDL Number MFCD00039159
UN Number UN3082
Solubility Information Insoluble in water
Health Hazard 2 GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 1 Warning
Beilstein 1858096
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
CAS 586-61-8

Applications
It employed as a intermediate for pharmaceutical.

Solubility
Insoluble in water

Notes
Store in cool, dry place in tightly closed container. Store away from oxidizing agent.

Safety and Handling

Warning

GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.