Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Bromo-4-fluoro-2-nitrobenzene, 98%


Quantity 1g

Manufacturer: alfa aesar™  L1550303

 View more versions of this product

Catalog No. AAL1550303

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 220.01
Chemical Name or Material 1-Bromo-4-fluoro-2-nitrobenzene
MDL Number MFCD00055530
Molecular Formula C6H3BrFNO2
Health Hazard 2 GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
CAS 446-09-3
UN Number UN2811
Synonym 2-Bromo-5-fluoronitrobenzene
Beilstein 2364227
Quantity 1g
Boiling Point 148° to 150°C (35mmHg)
Health Hazard 1 Danger
Assay 98%
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
Melting Point 39° to 42°C

Specifications

Chemical Name or Material 1-Bromo-4-fluoro-2-nitrobenzene
CAS 446-09-3
Molecular Formula C6H3BrFNO2
Formula Weight 220.01
MDL Number MFCD00055530
UN Number UN2811
Beilstein 2364227
Synonym 2-Bromo-5-fluoronitrobenzene
Accessories

Danger

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

SDS & Safety Documents