missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Bromo-4-difluoromethoxy-2-fluorobenzene, 96%

Chemical Identifiers

CAS 1242258-38-3
Molecular Formula C7H4BrF3O
Molecular Weight (g/mol) 241.007
MDL Number MFCD18390897
InChI Key QWUYTYZPBMAYKP-UHFFFAOYSA-N
Synonym 1-bromo-4-difluoromethoxy-2-fluorobenzene, benzene,1-bromo-4-difluoromethoxy-2-fluoro, 1-bromo-4-difluoromethoxy-2-fluoro-benzene, 1-bromo-4-difluoromethoxy-2-fluoro-benzene
PubChem CID 65203213
IUPAC Name 1-bromo-4-(difluoromethoxy)-2-fluorobenzene
SMILES C1=CC(=C(C=C1OC(F)F)F)Br
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH59931MD View Documents Alfa Aesar™
H59931MD
250mg N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AAH5993103 View Documents Alfa Aesar™
H5993103
1g N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

1242258-38-3
241.007
QWUYTYZPBMAYKP-UHFFFAOYSA-N
65203213
C1=CC(=C(C=C1OC(F)F)F)Br
C7H4BrF3O
MFCD18390897
1-bromo-4-difluoromethoxy-2-fluorobenzene, benzene,1-bromo-4-difluoromethoxy-2-fluoro, 1-bromo-4-difluoromethoxy-2-fluoro-benzene, 1-bromo-4-difluoromethoxy-2-fluoro-benzene
1-bromo-4-(difluoromethoxy)-2-fluorobenzene

Specifications

250mg
C7H4BrF3O
1-bromo-4-difluoromethoxy-2-fluorobenzene, benzene,1-bromo-4-difluoromethoxy-2-fluoro, 1-bromo-4-difluoromethoxy-2-fluoro-benzene, 1-bromo-4-difluoromethoxy-2-fluoro-benzene
C1=CC(=C(C=C1OC(F)F)F)Br
241.007
241.01
1-Bromo-4-difluoromethoxy-2-fluorobenzene
1242258-38-3
MFCD18390897
QWUYTYZPBMAYKP-UHFFFAOYSA-N
1-bromo-4-(difluoromethoxy)-2-fluorobenzene
65203213
96%
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning