missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
1-Bromo-4-decylbenzene 88.0+%, TCI America™
Supplier: TCI America B16115G
Specifications
1-Bromo-4-decylbenzene | |
Yellow | |
MFCD00061112 | |
1-bromo-4-n-decylbenzene, benzene, 1-bromo-4-decyl, 4-decylbromobenzene, acmc-2098jm, ksc909c2n | |
CCCCCCCCCCC1=CC=C(Br)C=C1 | |
297.28 | |
297.28 | |
Liquid |
106418-67-1 | |
C16H25Br | |
5 g | |
SWTGVSCSSDYINB-UHFFFAOYSA-N | |
1-bromo-4-decylbenzene | |
13757384 | |
≥88.0% (GC) |
Chemical Identifiers
106418-67-1 | |
297.28 | |
SWTGVSCSSDYINB-UHFFFAOYSA-N | |
13757384 | |
CCCCCCCCCCC1=CC=C(Br)C=C1 |
C16H25Br | |
MFCD00061112 | |
1-bromo-4-n-decylbenzene, benzene, 1-bromo-4-decyl, 4-decylbromobenzene, acmc-2098jm, ksc909c2n | |
1-bromo-4-decylbenzene |
Safety and Handling
TSCA : No