missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Bromo-4-chloro-2-nitrobenzene, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1263514

 View more versions of this product

Catalog No. AAA1263514


Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

1-Bromo-4-chloro-2-nitrobenzene
68°C to 72°C
MFCD00024320
1950424
UKTIMFAJRPSNGR-UHFFFAOYSA-N
1-bromo-4-chloro-2-nitrobenzene
2794904
98%
41513-04-6
C6H3BrClNO2
25 g
2-bromo-5-chloronitrobenzene, benzene, 1-bromo-4-chloro-2-nitro, 4-bromo-3-nitrochlorobenzene, 3-chloro-6-bromonitrobenzene, pubchem3593, acmc-209jkn, 1-bromo-4-chloronitrobenzene, ksc490m7r, 2-bromo-5-chloro-nitrobenzene, bromo-4-chloro-2-nitrobenzene
C1=CC(=C(C=C1Cl)[N+](=O)[O-])Br
236.449
236.45

Chemical Identifiers

41513-04-6
236.449
UKTIMFAJRPSNGR-UHFFFAOYSA-N
2794904
C1=CC(=C(C=C1Cl)[N+](=O)[O-])Br
C6H3BrClNO2
MFCD00024320
2-bromo-5-chloronitrobenzene, benzene, 1-bromo-4-chloro-2-nitro, 4-bromo-3-nitrochlorobenzene, 3-chloro-6-bromonitrobenzene, pubchem3593, acmc-209jkn, 1-bromo-4-chloronitrobenzene, ksc490m7r, 2-bromo-5-chloro-nitrobenzene, bromo-4-chloro-2-nitrobenzene
1-bromo-4-chloro-2-nitrobenzene
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

EINECSNumber : 255-421-4

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only