missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Bromo-4-chloro-2-fluorobenzene, TCI America™

$59.58 - $178.75

Chemical Identifiers

CAS 1996-29-8
Molecular Formula C6H3BrClF
Molecular Weight (g/mol) 209.442
InChI Key FPNVMCMDWZNTEU-UHFFFAOYSA-N
Synonym 4-chloro-2-fluorobromobenzene, 2-fluoro-4-chlorobromobenzene, 2-bromo-5-chlorofluorobenzene, 1-bromo-4-chloro-2-fluoro-benzene, 1-bromo-2-fluoro-4-chlorobenzene, 2-bromo-5-chloro-1-fluorobenzene, benzene, 1-bromo-4-chloro-2-fluoro, pubchem3209, acmc-1bovk, 2-bromo-5-chlorofluorbenzene
PubChem CID 137275
IUPAC Name 1-bromo-4-chloro-2-fluorobenzene
SMILES C1=CC(=C(C=C1Cl)F)Br
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B27245G View Documents TCI America
B27245G
5g Each for $59.58
 
B272425G View Documents TCI America
B272425G
25g Each for $178.75
 
Specifications

Chemical Identifiers

1996-29-8
209.442
4-chloro-2-fluorobromobenzene, 2-fluoro-4-chlorobromobenzene, 2-bromo-5-chlorofluorobenzene, 1-bromo-4-chloro-2-fluoro-benzene, 1-bromo-2-fluoro-4-chlorobenzene, 2-bromo-5-chloro-1-fluorobenzene, benzene, 1-bromo-4-chloro-2-fluoro, pubchem3209, acmc-1bovk, 2-bromo-5-chlorofluorbenzene
1-bromo-4-chloro-2-fluorobenzene
C6H3BrClF
FPNVMCMDWZNTEU-UHFFFAOYSA-N
137275
C1=CC(=C(C=C1Cl)F)Br

Specifications

92°C
5g
C6H3BrClF
FPNVMCMDWZNTEU-UHFFFAOYSA-N
1-bromo-4-chloro-2-fluorobenzene
137275
Clear Liquid at 20°C
1-Bromo-4-chloro-2-fluorobenzene
Yellow
1996-29-8
4-chloro-2-fluorobromobenzene, 2-fluoro-4-chlorobromobenzene, 2-bromo-5-chlorofluorobenzene, 1-bromo-4-chloro-2-fluoro-benzene, 1-bromo-2-fluoro-4-chlorobenzene, 2-bromo-5-chloro-1-fluorobenzene, benzene, 1-bromo-4-chloro-2-fluoro, pubchem3209, acmc-1bovk, 2-bromo-5-chlorofluorbenzene
C1=CC(=C(C=C1Cl)F)Br
209.442
209.44
≥98.0% (GC)
Safety and Handling

Safety and Handling

TSCA : No