missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Bromo-4-chloro-2-fluorobenzene, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC294050050

 View more versions of this product

Catalog No. AC294050050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

1-Bromo-4-chloro-2-fluorobenzene
Glass bottle
Colorless to Yellow
97%
96.0
97%
MFCD00079708
Solubility in water: insoluble in water. Other solubilities: soluble in most organic solvents
C1=CC(=C(C=C1Cl)F)Br
209.44
209.44
97%
Authentic
1.5540 to 1.5560
5g
1996-29-8
100.0
C6H3BrClF
1-bromo-2-fluoro-4-chlorobenzene, 1-bromo-4-chloro-2-fluoro-benzene, 2-bromo-5-chloro-1-fluorobenzene, 2-bromo-5-chlorofluorbenzene, 2-bromo-5-chlorofluorobenzene, 2-fluoro-4-chlorobromobenzene, 4-chloro-2-fluorobromobenzene, acmc-1bovk, benzene, 1-bromo-4-chloro-2-fluoro, pubchem3209
FPNVMCMDWZNTEU-UHFFFAOYSA-N
1-bromo-4-chloro-2-fluorobenzene
137275
Liquid

Chemical Identifiers

1996-29-8
209.44
FPNVMCMDWZNTEU-UHFFFAOYSA-N
137275
C1=CC(=C(C=C1Cl)F)Br
C6H3BrClF
MFCD00079708
1-bromo-2-fluoro-4-chlorobenzene, 1-bromo-4-chloro-2-fluoro-benzene, 2-bromo-5-chloro-1-fluorobenzene, 2-bromo-5-chlorofluorbenzene, 2-bromo-5-chlorofluorobenzene, 2-fluoro-4-chlorobromobenzene, 4-chloro-2-fluorobromobenzene, acmc-1bovk, benzene, 1-bromo-4-chloro-2-fluoro, pubchem3209
1-bromo-4-chloro-2-fluorobenzene
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS Documents