missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Bromo-4-chloro-2-fluorobenzene, 97%, ACROS Organics™

Chemical Identifiers

CAS 1996-29-8
Molecular Formula C6H3BrClF
Molecular Weight (g/mol) 209.44
MDL Number MFCD00079708
InChI Key FPNVMCMDWZNTEU-UHFFFAOYSA-N
Synonym 1-bromo-2-fluoro-4-chlorobenzene
PubChem CID 137275
IUPAC Name 1-bromo-4-chloro-2-fluorobenzene
SMILES C1=CC(=C(C=C1Cl)F)Br
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC294050050 SDS View Documents Thermo Fisher Scientific
AC294050050
5g Glass bottle N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

1996-29-8
209.44
FPNVMCMDWZNTEU-UHFFFAOYSA-N
137275
C1=CC(=C(C=C1Cl)F)Br
C6H3BrClF
MFCD00079708
1-bromo-2-fluoro-4-chlorobenzene
1-bromo-4-chloro-2-fluorobenzene

Specifications

Authentic
1.5540 to 1.5560
5g
96.0
97%
MFCD00079708
Solubility in water: insoluble in water. Other solubilities: soluble in most organic solvents
C1=CC(=C(C=C1Cl)F)Br
209.44
209.44
97%
Glass bottle
Colorless to Yellow
1996-29-8
100.0
C6H3BrClF
1-bromo-2-fluoro-4-chlorobenzene
FPNVMCMDWZNTEU-UHFFFAOYSA-N
1-bromo-4-chloro-2-fluorobenzene
137275
Liquid
1-Bromo-4-chloro-2-fluorobenzene, 97%
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning