missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Bromo-4-butylbenzene, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC320442500

 View more versions of this product

Catalog No. AC320442500

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

≥96.0%
213.12
Clear colorless to yellow
96.0
BrC6H4(CH)3CH3
1.5300 to 1.5330 (20°C, 589nm)
41492-05-1
MFCD00040934
Authentic
Liquid
250g
1.2g/mL
100.0
1-Bromo-4-butylbenzene
C10H13Br
97%
Glass bottle

Chemical Identifiers

41492-05-1
100.0
C10H13Br
213.12
1-Bromo-4-butylbenzene
96.0
97%
BrC6H4(CH)3CH3
MFCD00040934
≥96.0%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.