Please login to your online account to display your discounted pricing

1-Bromo-4-butylbenzene, 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Ethyl bromofluoroacetate
Name Note 97%
CAS 401-55-8
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C4H6BrFO2
Linear Formula BrCH(F)CO2C2H5
Formula Weight 185
MDL Number MFCD00042095
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC320480010

  SDS
thermo fisher scientific
AC320480010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥96.0%
Formula Weight 185
Physical Form Liquid
Quantity 1g
Color Clear yellow
Density 1.5650g/mL
EINECS Number 206-928-4
Refractive Index 1.4240 to 1.4280 (20°C, 589nm)
Infrared Spectrum Authentic
Molecular Formula C4H6BrFO2
Packaging Glass bottle
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Harmful in contact with skin.
Harmful if inhaled.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Name Note 97%
Health Hazard 1 GHS Signal Word: Warning
Assay Percent Range 97%
Linear Formula BrCH(F)CO2C2H5
MDL Number MFCD00042095
CAS 401-55-8
Chemical Name or Material Ethyl bromofluoroacetate
Safety and Handling

GHS Signal Word: Warning

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Harmful in contact with skin.
Harmful if inhaled.
Causes serious eye irritation.
May cause respiratory irritation.