missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
1-Bromo-4-butoxybenzene, 97%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals BTB09793DE
Specifications
1-Bromo-4-butoxybenzene | |
96.5 | |
97% | |
BrC6H4O(CH2)3CH3 | |
5g | |
BOUVKHWPQNEXTO-UHFFFAOYSA-N | |
1-bromo-4-butoxybenzene | |
142395 | |
97% |
39969-57-8 | |
100.0 | |
C10H13BrO | |
MFCD00037100 | |
p-bromophenyl butyl ether, benzene,1-bromo-4-butoxy, p-butoxybromobenzene, benzene, 1-bromo-4-butoxy, 4-bromo-1-butoxybenzene, 4-butoxybromobenzene, 4-n-butoxyphenyl bromide, 1-butoxy-4-bromobenzene, 4-bromophenyl butyl ether | |
CCCCOC1=CC=C(C=C1)Br | |
229.117 | |
229.12 |