missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Bromo-4,5-dichloro-2-fluorobenzene, 96%

$137.39 - $462.64

Chemical Identifiers

CAS 1000572-78-0
Molecular Formula C6H2BrCl2F
Molecular Weight (g/mol) 243.884
MDL Number MFCD09878158
InChI Key YAKOUNMAKAPKIQ-UHFFFAOYSA-N
Synonym 4,5-dichloro-2-fluorobromobenzene, benzene,1-bromo-4,5-dichloro-2-fluoro
PubChem CID 44891196
IUPAC Name 1-bromo-4,5-dichloro-2-fluorobenzene
SMILES C1=C(C(=CC(=C1Cl)Cl)Br)F
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH59984MD View Documents Alfa Aesar™
H59984MD
250mg Each for $137.39
 
AAH5998403 View Documents Alfa Aesar™
H5998403
1g Each for $462.64
 
Specifications

Chemical Identifiers

1000572-78-0
243.884
YAKOUNMAKAPKIQ-UHFFFAOYSA-N
44891196
C1=C(C(=CC(=C1Cl)Cl)Br)F
C6H2BrCl2F
MFCD09878158
4,5-dichloro-2-fluorobromobenzene, benzene,1-bromo-4,5-dichloro-2-fluoro
1-bromo-4,5-dichloro-2-fluorobenzene

Specifications

250mg
C6H2BrCl2F
4,5-dichloro-2-fluorobromobenzene, benzene,1-bromo-4,5-dichloro-2-fluoro
C1=C(C(=CC(=C1Cl)Cl)Br)F
243.884
243.89
1-Bromo-4,5-dichloro-2-fluorobenzene
1000572-78-0
MFCD09878158
YAKOUNMAKAPKIQ-UHFFFAOYSA-N
1-bromo-4,5-dichloro-2-fluorobenzene
44891196
96%
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning