Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Bromo-4,4,4-trifluorobutane, 98%

$121.74 - $470.11

Chemical Identifiers

Chemical Name or Material 1-Bromo-4,4,4-trifluorobutane
CAS 406-81-5
Molecular Formula CF3CH2CH2CH2Br
Formula Weight 191
MDL Number MFCD00066335
Beilstein 1736058
Synonym 4,4,4-Trifluoro-1-bromobutane; 4,4,4-Trifluorobutyl bromide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1023703

  SDS
alfa aesar™
L1023703
1g Each for $121.74

AAL1023706

  SDS
alfa aesar™
L1023706
5g Each for $470.11
Description & Specifications

Specifications

Formula Weight 191
CAS 406-81-5
Chemical Name or Material 1-Bromo-4,4,4-trifluorobutane
Molecular Formula CF3CH2CH2CH2Br
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Beilstein 1736058
Assay 98%
Boiling Point 103° to 105°C
MDL Number MFCD00066335
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 1 Warning
Synonym 4,4,4-Trifluoro-1-bromobutane; 4,4,4-Trifluorobutyl bromide
Quantity 1g

Applications
1-Bromo-4,4,4-trifluorobutane is used as pharmaceutical intermediate.

Solubility
Not miscible in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.