Please login to your online account to display your discounted pricing

1-Bromo-3-phenylpropane, 98%, ACROS Organics™


Quantity 100mL
Packaging Glass bottle

Manufacturer: fisher scientific acros  107291000

 View more versions of this product

Catalog No. AC107291000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Liquid
Formula Weight 199.09
Assay ≥97.5%
Quantity 100mL
Color Clear colorless to light yellow
Beilstein 05, 391
TSCA TSCA
EINECS Number 211-294-7
CAS Max % 100.0
Synonym (3-Bromopropyl)benzene
Packaging Glass bottle
Refractive Index 1.5450 to 1.5470 (20°C, 589nm)
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Molecular Formula C9H11Br
Assay Percent Range 98%
Linear Formula C6H5(CH2)3Br
MDL Number MFCD00000257
Chemical Name or Material 1-Bromo-3-phenylpropane
CAS Min % 97.5
Name Note 98%
CAS 637-59-2
Infrared Spectrum Authentic
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material 1-Bromo-3-phenylpropane
Name Note 98%
CAS 637-59-2
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C9H11Br
Linear Formula C6H5(CH2)3Br
Formula Weight 199.09
MDL Number MFCD00000257
Synonym (3-Bromopropyl)benzene
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.

SDS & Safety Documents