Learn More
1-Bromo-3-phenylpropane, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1405036
Description
1-Bromo-3-phenylpropane is used in the synthesis of MraY natural inhibitors as antibacterial agents. Also used in the preparation of quinolinone derivatives as potent and selective MAO-B inhibitors.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1-Bromo-3-phenylpropane is used in the synthesis of MraY natural inhibitors as antibacterial agents. Also used in the preparation of quinolinone derivatives as potent and selective MAO-B inhibitors.
Solubility
Soluble in water (partly). Soluble in chloroform and methanol.
Notes
Store in cool, dry place in tightly closed containers. Store away from oxidizing agent.
Specifications
1-Bromo-3-phenylpropane | |
1.308 | |
101°C (213°F) | |
1.546 | |
500 g | |
3-bromopropyl benzene, 1-bromo-3-phenylpropane, 3-phenylpropyl bromide, benzene, 3-bromopropyl, 3-bromo-1-phenylpropane, 3-bromoprop-1-yl benzene, unii-6vx623qn9v, .gamma.-bromopropyl benzene, 4-bromopropylbenzene, benzene, bromopropyl | |
XMZQWZJMTBCUFT-UHFFFAOYSA-N | |
3-bromopropylbenzene | |
12503 | |
98% |
637-59-2 | |
219°C to 220°C | |
C9H11Br | |
MFCD00000257 | |
2205527 | |
Soluble in water (partly). Soluble in chloroform and methanol. | |
BrCCCC1=CC=CC=C1 | |
199.09 | |
199.09 |
Chemical Identifiers
637-59-2 | |
199.09 | |
XMZQWZJMTBCUFT-UHFFFAOYSA-N | |
12503 | |
BrCCCC1=CC=CC=C1 |
C9H11Br | |
MFCD00000257 | |
3-bromopropyl benzene, 1-bromo-3-phenylpropane, 3-phenylpropyl bromide, benzene, 3-bromopropyl, 3-bromo-1-phenylpropane, 3-bromoprop-1-yl benzene, unii-6vx623qn9v, .gamma.-bromopropyl benzene, 4-bromopropylbenzene, benzene, bromopropyl | |
3-bromopropylbenzene |
Safety and Handling
GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 211-294-7
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only