Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Bromo-3-n-hexylbenzene, 97+%

$140.39 - $418.69

Chemical Identifiers

Chemical Name or Material 1-Bromo-3-n-hexylbenzene
CAS 38409-59-5
Molecular Formula C12H17Br
Formula Weight 241.17
MDL Number MFCD09909567
Synonym 3-(n-Hexyl)bromobenzene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2606306

 
alfa aesar™
H2606306
5g Each for $140.39

AAH2606314

 
alfa aesar™
H2606314
25g Each for $418.69
Description & Specifications

Specifications

Formula Weight 241.17
Chemical Name or Material 1-Bromo-3-n-hexylbenzene
Boiling Point 90° to 98°C (5mmHg)
Quantity 5g
Flash Point >120°C (248°F)
Assay ≥97%
Molecular Formula C12H17Br
MDL Number MFCD09909567
CAS 38409-59-5
Synonym 3-(n-Hexyl)bromobenzene
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Applications
1-Bromo-3-n-hexylbenzene is used as intermediates in pharmaceutical industry.

Solubility
Insoluble in water. Fully miscible with ethanol, acetone, toluene, methylene chloride.

Notes
Store in a cool, dry place in tightly closed container. Incompatible with oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.