missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Bromo-3-methylbutane, 99%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 403055000

 View more versions of this product

Catalog No. AC403055000

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

0.1% max. (as HBr)
99%
98.5
Authentic
1.4399 to 1.4419 (20°C, 589nm)
1.2000g/mL
Clear colorless
≥98.5%
C5H11Br
151.05
01, 136
Isoamyl bromide
1-Bromo-3-methylbutane
107-82-4
100.0
Glass bottle
0.05% max. (K.F.)
500g
Liquid
99%
(CH3)2CHCH2CH2Br
MFCD00000253
15, 5159

Chemical Identifiers

107-82-4
100.0
C5H11Br
151.05
Isoamyl bromide
≥98.5%
98.5
99%
(CH3)2CHCH2CH2Br
MFCD00000253
1-Bromo-3-methylbutane, 99%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Toxic to aquatic life with long lasting effects.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Flammable liquid and vapor.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning

EINECSNumber : 203-522-9

RTECSNumber : EJ6230000

TSCA : TSCA