missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Bromo-3-isopropylbenzene, 95%

$62.00 - $205.11

Chemical Identifiers

CAS 5436-01-1
Molecular Formula C5H6N2O2
Molecular Weight (g/mol) 126.12
MDL Number MFCD01318112
InChI Key UAECOHJYXUJDOF-UHFFFAOYSA-N
Synonym 3-hydroxy-1-methylpyridazin-6 1h-one, 1-methyl-3,6-pyridazinedione, n-methyl-3,6-pyridazinedione, n-methylmaleic hydrazide, 6-hydroxy-2-methylpyridazin-3 2h-one, n-methyl maleic hydrazide, 1-methyl-3,6-1h,2h pyridazinedione, 1-methyl-1,2-dihydropyridazine-3,6-dione, 6-hydroxy-2-methylpyridazin-3-one, 1-methyl-1,2-dihydro-3,6-pyridazinedione
PubChem CID 95157
IUPAC Name 2-methyl-1H-pyridazine-3,6-dione
SMILES CN1C(=O)C=CC(=O)N1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1765403 View Documents Alfa Aesar™
L1765403
1g Each for $62.00
 
AAL1765406 View Documents Alfa Aesar™
L1765406
5g Each for $205.11
 
Description

Description

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

Solubility
Difficult to mix in water.

Notes
Store in cool dry conditions in well sealed containers. Incompatible with strong oxidizing agents.
Specifications

Chemical Identifiers

5436-01-1
126.12
UAECOHJYXUJDOF-UHFFFAOYSA-N
95157
CN1C(=O)C=CC(=O)N1
C5H6N2O2
MFCD01318112
3-hydroxy-1-methylpyridazin-6 1h-one, 1-methyl-3,6-pyridazinedione, n-methyl-3,6-pyridazinedione, n-methylmaleic hydrazide, 6-hydroxy-2-methylpyridazin-3 2h-one, n-methyl maleic hydrazide, 1-methyl-3,6-1h,2h pyridazinedione, 1-methyl-1,2-dihydropyridazine-3,6-dione, 6-hydroxy-2-methylpyridazin-3-one, 1-methyl-1,2-dihydro-3,6-pyridazinedione
2-methyl-1H-pyridazine-3,6-dione

Specifications

210°C
5433-01-2
MFCD01318112
1-bromo-3-isopropylbenzene, 3-isopropylbromobenzene, 3-bromocumene, 3-bromoisopropylbenzene, benzene, 1-bromo-3-1-methylethyl, 1-bromo-3-isopropyl-benzene, 1-bromo-3-1-methylethyl benzene, m-bromocumene, m-isopropylbromobenzene, m-bromo-isopropyl-benzene
CC(C)C1=CC(=CC=C1)Br
199.09
199.1
1-Bromo-3-isopropylbenzene
1g
C9H11Br
1858092
GBSGGFCCQZUXNB-UHFFFAOYSA-N
1-bromo-3-propan-2-ylbenzene
138499
95%
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.

Warning