missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Bromo-3-isopropoxybenzene, 98%

$45.93 - $141.07

Chemical Identifiers

CAS 131738-73-3
Molecular Formula C9H11BrO
Formula Weight 215.09
MDL Number MFCD00070756
Beilstein 4176950
Synonym 2-(3-Bromophenoxy)propane; 3-Bromophenyl isopropyl ether
Chemical Name or Material 1-Bromo-3-isopropoxybenzene
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1380206 SDS Alfa Aesar™
L1380206
5g Each for $45.93
AAL1380214 SDS Alfa Aesar™
L1380214
25g Each for $141.07
Specifications Description & Specifications

Chemical Identifiers

131738-73-3
215.09
4176950
1-Bromo-3-isopropoxybenzene
C9H11BrO
MFCD00070756
2-(3-Bromophenoxy)propane; 3-Bromophenyl isopropyl ether

Specifications

131738-73-3
215.09
4176950
222°C
108°C (226°F)
98%
C9H11BrO
MFCD00070756
2-(3-Bromophenoxy)propane; 3-Bromophenyl isopropyl ether
5g
1-Bromo-3-isopropoxybenzene