Alfa Aesar™ 1-Bromo-3-isopropoxybenzene, 98%
Manufacturer:  Alfa Aesar™ L1380206
 View more versions of this product
Catalog No. AAL1380206
Specifications
Specifications
1-Bromo-3-isopropoxybenzene
222°C
131738-73-3
MFCD00070756
1-bromo-3-isopropoxybenzene, 3-bromophenyl isopropyl ether, 1-bromo-3-propan-2-yloxy benzene, 3-bromo-isopropoxybenzene, 3-bromoisoproproxylbenzene, 2-3-bromophenoxy propane, 3-bromoisopropoxybenzene, 3-bromo phenyl isopropyl ether, benzene, 1-bromo-3-1-methylethoxy, pubchem14867
CC(C)OC1=CC(=CC=C1)Br
215.09
215.09
108°C (226°F)
5g
C9H11BrO
4176950
JYWJZIQSPRFCDB-UHFFFAOYSA-N
1-bromo-3-propan-2-yloxybenzene
3260801
98%
Chemical Identifiers
131738-73-3
215.09
JYWJZIQSPRFCDB-UHFFFAOYSA-N
3260801
CC(C)OC1=CC(=CC=C1)Br
C9H11BrO
MFCD00070756
1-bromo-3-isopropoxybenzene, 3-bromophenyl isopropyl ether, 1-bromo-3-propan-2-yloxy benzene, 3-bromo-isopropoxybenzene, 3-bromoisoproproxylbenzene, 2-3-bromophenoxy propane, 3-bromoisopropoxybenzene, 3-bromo phenyl isopropyl ether, benzene, 1-bromo-3-1-methylethoxy, pubchem14867
1-bromo-3-propan-2-yloxybenzene
Documents