missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Bromo-3-iodobenzene, 98+%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC249420500

 View more versions of this product

Catalog No. AC249420500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

1-Bromo-3-iodobenzene
>110°C
1.6590 to 1.6615
120.0°C (18.0 mmHg)
50mL
98.0
98% min. (GC)
BrC6H4I
05, 223
Solubility in water: insoluble. Other solubilities: soluble in most common organic solvents
C1=CC(=CC(=C1)I)Br
282.89
282.89
2.2100g/mL
Glass bottle
2.21
-9.3°C
591-18-4
100.0
C6H4BrI
MFCD00001043
1-bromo-3-iodo-benzene, 3-bromo iodobenzene, 3-bromo-1-iodobenzene, 3-bromoiodobenzene, 3-iodobromobenzene, attercop-chm at127411, benzene, 1-bromo-3-iodo, m-bromjodbenzol, m-bromoiodobenzene
CTPUUDQIXKUAMO-UHFFFAOYSA-N
1-bromo-3-iodobenzene
11561
98+%

Chemical Identifiers

591-18-4
282.89
CTPUUDQIXKUAMO-UHFFFAOYSA-N
11561
C1=CC(=CC(=C1)I)Br
C6H4BrI
MFCD00001043
1-bromo-3-iodo-benzene, 3-bromo iodobenzene, 3-bromo-1-iodobenzene, 3-bromoiodobenzene, 3-iodobromobenzene, attercop-chm at127411, benzene, 1-bromo-3-iodo, m-bromjodbenzol, m-bromoiodobenzene
1-bromo-3-iodobenzene
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 209-703-9

SDS Documents