Learn More
1-Bromo-3-iodobenzene, 98+%, Thermo Scientific™
Chemical Identifiers
CAS | 591-18-4 |
---|---|
Molecular Formula | C6H4BrI |
Molecular Weight (g/mol) | 282.91 |
MDL Number | MFCD00001043 |
InChI Key | CTPUUDQIXKUAMO-UHFFFAOYSA-N |
Synonym | 3-bromoiodobenzene, benzene, 1-bromo-3-iodo, m-bromoiodobenzene, 1-bromo-3-iodo-benzene, 3-bromo-1-iodobenzene, attercop-chm at127411, m-bromjodbenzol, 3-iodobromobenzene, 3-bromo iodobenzene |
PubChem CID | 11561 |
IUPAC Name | 1-bromo-3-iodobenzene |
SMILES | BrC1=CC=CC(I)=C1 |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC249420100
|
Thermo Scientific Chemicals
249420100 |
10mL | Glass bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
AC249420500
|
Thermo Scientific Chemicals
249420500 |
50mL | Glass bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
591-18-4 | |
282.91 | |
CTPUUDQIXKUAMO-UHFFFAOYSA-N | |
11561 | |
BrC1=CC=CC(I)=C1 |
C6H4BrI | |
MFCD00001043 | |
3-bromoiodobenzene, benzene, 1-bromo-3-iodo, m-bromoiodobenzene, 1-bromo-3-iodo-benzene, 3-bromo-1-iodobenzene, attercop-chm at127411, m-bromjodbenzol, 3-iodobromobenzene, 3-bromo iodobenzene | |
1-bromo-3-iodobenzene |
Specifications
591-18-4 | |
100.0 | |
120.0°C (18.0 mmHg) | |
98% min. (GC) | |
C6H4BrI | |
BrC6H4I | |
10mL | |
2.21 | |
Solubility in water: insoluble. Other solubilities: soluble in most common organic solvents | |
BrC1=CC=CC(I)=C1 | |
282.91 | |
282.89 | |
1-Bromo-3-iodobenzene |
98.0 | |
2.2100g/mL | |
>110°C | |
Glass bottle | |
1.6590 to 1.6615 | |
MFCD00001043 | |
05, 223 | |
3-bromoiodobenzene, benzene, 1-bromo-3-iodo, m-bromoiodobenzene, 1-bromo-3-iodo-benzene, 3-bromo-1-iodobenzene, attercop-chm at127411, m-bromjodbenzol, 3-iodobromobenzene, 3-bromo iodobenzene | |
CTPUUDQIXKUAMO-UHFFFAOYSA-N | |
1-bromo-3-iodobenzene | |
11561 | |
98+% |
Safety and Handling
GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
EINECSNumber : 209-703-9
RUO â Research Use Only