Learn More
1-Bromo-3-fluorobenzene, 99%, Thermo Scientific Chemicals
$90.75 - $589.64
Chemical Identifiers
CAS | 1073-06-9 |
---|---|
Molecular Formula | C6H4BrF |
MDL Number | MFCD00000326 |
InChI Key | QDFKKJYEIFBEFC-UHFFFAOYSA-N |
Synonym | 3-bromofluorobenzene, m-bromofluorobenzene, m-fluorobromobenzene, benzene, 1-bromo-3-fluoro, 3-bromfluorbenzen, 3-fluorobromobenzene, m-fluorophenyl bromide, 1-fluoro-3-bromobenzene, 3-fluoro bromobenzene, benzene, 1-fluoro-3-bromo |
PubChem CID | 14082 |
IUPAC Name | 1-bromo-3-fluorobenzene |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1085814
|
Thermo Scientific Chemicals
A1085814 |
25 g |
Each for $90.75
|
|
|||||
AAA1085818
|
Thermo Scientific Chemicals
A1085818 |
50 g |
Each for $141.19
|
|
|||||
AAA1085822
|
Thermo Scientific Chemicals
A1085822 |
100 g |
Each for $248.75
|
|
|||||
AAA1085830
|
Thermo Scientific Chemicals
A1085830 |
250 g |
Each for $589.64
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1073-06-9 | |
MFCD00000326 | |
3-bromofluorobenzene, m-bromofluorobenzene, m-fluorobromobenzene, benzene, 1-bromo-3-fluoro, 3-bromfluorbenzen, 3-fluorobromobenzene, m-fluorophenyl bromide, 1-fluoro-3-bromobenzene, 3-fluoro bromobenzene, benzene, 1-fluoro-3-bromo | |
1-bromo-3-fluorobenzene |
C6H4BrF | |
QDFKKJYEIFBEFC-UHFFFAOYSA-N | |
14082 |
Specifications
1073-06-9 | |
1.594 | |
46°C (115°F) | |
C6H4BrF | |
MFCD00000326 | |
UN1993 | |
3-bromofluorobenzene, m-bromofluorobenzene, m-fluorobromobenzene, benzene, 1-bromo-3-fluoro, 3-bromfluorbenzen, 3-fluorobromobenzene, m-fluorophenyl bromide, 1-fluoro-3-bromobenzene, 3-fluoro bromobenzene, benzene, 1-fluoro-3-bromo | |
C1=CC(=CC(=C1)Br)F | |
175 | |
175.01 | |
1-Bromo-3-fluorobenzene |
-8°C | |
148°C to 150°C | |
Aromatic | |
1.5275 | |
25 g | |
1853969 | |
QDFKKJYEIFBEFC-UHFFFAOYSA-N | |
1-bromo-3-fluorobenzene | |
14082 | |
99% |
Safety and Handling
GHS H Statement
H226-H302-H315-H319
Flammable liquid and vapor.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
H226-H315-H319-H335
DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber : 214-023-0
RTECSNumber : DA1225000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only