missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Bromo-3-fluoro-5-nitrobenzene, 98%

Manufacturer:  Alfa Aesar™ H6119206

 View more versions of this product

Catalog No. AAH6119206



Specifications

Specifications

1-Bromo-3-fluoro-5-nitrobenzene
5g
C6H3BrFNO2
UN2810
SWXVEPMSQBEVRH-UHFFFAOYSA-N
1-bromo-3-fluoro-5-nitrobenzene
15020157
Liquid
Yellow
7087-65-2
MFCD09955454
1-nitro-3-fluoro-5-bromobenzene
C1=C(C=C(C=C1F)Br)[N+](=O)[O-]
219.997
220
98%

Chemical Identifiers

7087-65-2
219.997
SWXVEPMSQBEVRH-UHFFFAOYSA-N
15020157
C1=C(C=C(C=C1F)Br)[N+](=O)[O-]
C6H3BrFNO2
MFCD09955454
1-nitro-3-fluoro-5-bromobenzene
1-bromo-3-fluoro-5-nitrobenzene
Safety and Handling

Safety and Handling

GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335

DOTInformation : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures

Documents