missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Bromo-3-fluoro-4-iodobenzene, 99+%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC249401000

 View more versions of this product

Catalog No. AC249401000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

1-Bromo-3-fluoro-4-iodobenzene
Authentic
White to Beige
100g
99.0
99% min. (GC)
BrC6H3(F)I
1-bromo-3-fluoro-4-iodobenzene, 2-fluoro-4-bromo iodobenzene, 2-fluoro-4-bromoiodobenzene, 3-fluoro-4-iodo-bromobenzene, 3-fluoro-4-iodobromobenzene, 4-bromo-2-fluoro-1-iodo-benzene, 4-bromo-2-fluoroiodobenzene, acmc-2098hz, benzene, 4-bromo-2-fluoro-1-iodo, pubchem3232
C1=CC(=C(C=C1Br)F)I
300.89
300.89
99+%
>110°C
Glass bottle
46°C to 49°C
105931-73-5
100.0
C6H3BrFI
MFCD00010608
XRMZKCQCINEBEI-UHFFFAOYSA-N
4-bromo-2-fluoro-1-iodobenzene
2724516
Crystalline Powder, Crystals, and/or Chunks

Chemical Identifiers

105931-73-5
300.89
XRMZKCQCINEBEI-UHFFFAOYSA-N
2724516
C1=CC(=C(C=C1Br)F)I
C6H3BrFI
MFCD00010608
1-bromo-3-fluoro-4-iodobenzene, 2-fluoro-4-bromo iodobenzene, 2-fluoro-4-bromoiodobenzene, 3-fluoro-4-iodo-bromobenzene, 3-fluoro-4-iodobromobenzene, 4-bromo-2-fluoro-1-iodo-benzene, 4-bromo-2-fluoroiodobenzene, acmc-2098hz, benzene, 4-bromo-2-fluoro-1-iodo, pubchem3232
4-bromo-2-fluoro-1-iodobenzene
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS Documents