Please login to your online account to display your discounted pricing

1-Bromo-3-ethylbenzene, 98%, Alfa Aesar™

$209.45 - $209.45

Chemical Identifiers

Chemical Name or Material 1-Bromo-3-ethylbenzene
CAS 2725-82-8
Molecular Formula C8H9Br
Formula Weight 185.06
MDL Number MFCD00156128
Synonym 3-Ethylbromobenzene
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AAA1225806

  SDS
alfa aesar
A1225806
Each for $209.45
Description & Specifications

Specifications

Formula Weight 185.06
Quantity 5g
Chemical Name or Material 1-Bromo-3-ethylbenzene
Assay 98%
Boiling Point 76° to 78°C (10mmHg)
Molecular Formula C8H9Br
MDL Number MFCD00156128
CAS 2725-82-8
Synonym 3-Ethylbromobenzene
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335;
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a;
Avoid breathing dust/fume/gas/mist/vapours/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a;
Avoid breathing dust/fume/gas/mist/vapours/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335;
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.