missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Bromo-3-ethylbenzene, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1225803

 View more versions of this product

Catalog No. AAA1225803



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

1-Bromo-3-ethylbenzene
1.3493
C8H9Br
1 g
ZRFJYAZQMFCUIX-UHFFFAOYSA-N
1-bromo-3-ethylbenzene
123170
98%
2725-82-8
76°C to 78°C (10 mmHg)
MFCD00156128
benzene, 1-bromo-3-ethyl, 3-ethylbromobenzene, 1-bromo-3-ethyl-benzene, 3-bromoethyl benzene, 3ethylbromobenzene, m-bromoethylbenzene, 3-bromo-ethylbenzene, pubchem3771, 1bromo-3-ethylbenzene, 1-bromo-3-ethylbenze
CCC1=CC(=CC=C1)Br
185.064
185.06

Chemical Identifiers

2725-82-8
185.064
ZRFJYAZQMFCUIX-UHFFFAOYSA-N
123170
CCC1=CC(=CC=C1)Br
C8H9Br
MFCD00156128
benzene, 1-bromo-3-ethyl, 3-ethylbromobenzene, 1-bromo-3-ethyl-benzene, 3-bromoethyl benzene, 3ethylbromobenzene, m-bromoethylbenzene, 3-bromo-ethylbenzene, pubchem3771, 1bromo-3-ethylbenzene, 1-bromo-3-ethylbenze
1-bromo-3-ethylbenzene
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only