Please login to your online account to display your discounted pricing

1-Bromo-3-ethylbenzene, 97%, Maybridge

Chemical Identifiers

Chemical Name or Material 1-Bromo-3-ethylbenzene
CAS 2725-82-8
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C8H9Br
Linear Formula CH3CH2C6H4Br
Formula Weight 185.06
MDL Number MFCD00156128
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

TL00571DA

 
fisher scientific company
TL00571DA
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 185.06
Quantity 1g
Assay 97%
CAS Min % 97.0
Chemical Name or Material 1-Bromo-3-ethylbenzene
Assay Percent Range 97%
Linear Formula CH3CH2C6H4Br
CAS 2725-82-8
MDL Number MFCD00156128
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Molecular Formula C8H9Br
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.