Please login to your online account to display your discounted pricing

1-Bromo-3-ethylbenzene, 97%, Maybridge


Quantity 1g

Manufacturer: fisher scientific company  TL00571DA

 View more versions of this product

Catalog No. TL00571DA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 185.06
Quantity 1g
Assay 97%
CAS Min % 97.0
Linear Formula CH3CH2C6H4Br
Assay Percent Range 97%
CAS 2725-82-8
Chemical Name or Material 1-Bromo-3-ethylbenzene
MDL Number MFCD00156128
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Molecular Formula C8H9Br
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material 1-Bromo-3-ethylbenzene
CAS 2725-82-8
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C8H9Br
Linear Formula CH3CH2C6H4Br
Formula Weight 185.06
MDL Number MFCD00156128
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

SDS & Safety Documents