Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Bromo-3-(difluoromethoxy)benzene, 97+%

$78.50 - $274.23

Chemical Identifiers

Chemical Name or Material 1-Bromo-3-(difluoromethoxy)benzene
CAS 262587-05-3
Molecular Formula C7H5BrF2O
Formula Weight 223.02
MDL Number MFCD00236233
Synonym 3-(Difluoromethoxy)bromobenzene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2397703

  SDS
alfa aesar™
B2397703
1g Each for $78.50

AAB2397706

  SDS
alfa aesar™
B2397706
5g Each for $274.23
Description & Specifications

Specifications

Formula Weight 223.02
MDL Number MFCD00236233
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Molecular Formula C7H5BrF2O
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
CAS 262587-05-3
Health Hazard 1 Warning
Quantity 1g
Synonym 3-(Difluoromethoxy)bromobenzene
Chemical Name or Material 1-Bromo-3-(difluoromethoxy)benzene
Assay ≥97%
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.