missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
1-Bromo-3-cyclopropylbenzene, 96%, ACROS Organics™
Chemical Identifiers
CAS | 1798-85-2 |
---|---|
Molecular Formula | C9H9Br |
Molecular Weight (g/mol) | 197.07 |
InChI Key | BCBNVQYWODLZSI-UHFFFAOYSA-N |
Synonym | 1-bromo-3-cyclopropyl benzene, 1-bromo-3-cyclopropyl-benzene, 1-cyclopropyl-3-bromobenzene, 3-bromo-1-cyclopropylbenzene, 3-cyclopropylbromobenzene, benzene, 1-bromo-3-cyclopropyl, benzene,1-bromo-3-cyclopropyl, bromo-3-cyclopropyl-benzene, m-bromophenylcyclopropane, pubchem22120 |
PubChem CID | 3282672 |
IUPAC Name | 1-bromo-3-cyclopropylbenzene |
SMILES | C1CC1C2=CC(=CC=C2)Br |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AC452582500
|
Thermo Fisher Scientific
AC452582500 |
250mg |
N/A
|
N/A | |||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
AC452580010
|
Thermo Fisher Scientific
AC452580010 |
1g |
N/A
|
N/A | |||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
1798-85-2 | |
197.07 | |
1-bromo-3-cyclopropyl benzene, 1-bromo-3-cyclopropyl-benzene, 1-cyclopropyl-3-bromobenzene, 3-bromo-1-cyclopropylbenzene, 3-cyclopropylbromobenzene, benzene, 1-bromo-3-cyclopropyl, benzene,1-bromo-3-cyclopropyl, bromo-3-cyclopropyl-benzene, m-bromophenylcyclopropane, pubchem22120 | |
1-bromo-3-cyclopropylbenzene |
C9H9Br | |
BCBNVQYWODLZSI-UHFFFAOYSA-N | |
3282672 | |
C1CC1C2=CC(=CC=C2)Br |
Specifications
250mg | |
95% min. (GC) | |
1-bromo-3-cyclopropyl benzene, 1-bromo-3-cyclopropyl-benzene, 1-cyclopropyl-3-bromobenzene, 3-bromo-1-cyclopropylbenzene, 3-cyclopropylbromobenzene, benzene, 1-bromo-3-cyclopropyl, benzene,1-bromo-3-cyclopropyl, bromo-3-cyclopropyl-benzene, m-bromophenylcyclopropane, pubchem22120 | |
C1CC1C2=CC(=CC=C2)Br | |
197.07 | |
197.07 | |
1-Bromo-3-cyclopropylbenzene |
1798-85-2 | |
C9H9Br | |
BCBNVQYWODLZSI-UHFFFAOYSA-N | |
1-bromo-3-cyclopropylbenzene | |
3282672 | |
96% |
Safety and Handling
GHS Signal Word: Warning