missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
1-Bromo-3-cyclopropylbenzene, 96%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC452582500
Specifications
1-Bromo-3-cyclopropylbenzene | |
1798-85-2 | |
C9H9Br | |
BCBNVQYWODLZSI-UHFFFAOYSA-N | |
1-bromo-3-cyclopropylbenzene | |
3282672 | |
96% |
250mg | |
95% min. (GC) | |
1-bromo-3-cyclopropyl benzene, 1-bromo-3-cyclopropyl-benzene, 1-cyclopropyl-3-bromobenzene, 3-bromo-1-cyclopropylbenzene, 3-cyclopropylbromobenzene, benzene, 1-bromo-3-cyclopropyl, benzene,1-bromo-3-cyclopropyl, bromo-3-cyclopropyl-benzene, m-bromophenylcyclopropane, pubchem22120 | |
C1CC1C2=CC(=CC=C2)Br | |
197.07 | |
197.07 |
Chemical Identifiers
1798-85-2 | |
197.07 | |
1-bromo-3-cyclopropyl benzene, 1-bromo-3-cyclopropyl-benzene, 1-cyclopropyl-3-bromobenzene, 3-bromo-1-cyclopropylbenzene, 3-cyclopropylbromobenzene, benzene, 1-bromo-3-cyclopropyl, benzene,1-bromo-3-cyclopropyl, bromo-3-cyclopropyl-benzene, m-bromophenylcyclopropane, pubchem22120 | |
1-bromo-3-cyclopropylbenzene |
C9H9Br | |
BCBNVQYWODLZSI-UHFFFAOYSA-N | |
3282672 | |
C1CC1C2=CC(=CC=C2)Br |
Safety and Handling
GHS Signal Word: Warning