Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Bromo-3-chloro-5-fluorobenzene, 98%

$103.76 - $1,402.28

Chemical Identifiers

Chemical Name or Material 1-Bromo-3-chloro-5-fluorobenzene
CAS 33863-76-2
Molecular Formula C6H3BrClF
Formula Weight 209.45
MDL Number MFCD00070747
Beilstein 2432912
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1659706

 
alfa aesar™
A1659706
5g Each for $103.76

AAA1659714

 
alfa aesar™
A1659714
25g Each for $415.61

AAA1659722

 
alfa aesar™
A1659722
100g Each for $1,402.28
Description & Specifications

Specifications

Formula Weight 209.45
Chemical Name or Material 1-Bromo-3-chloro-5-fluorobenzene
Assay 98%
Molecular Formula C6H3BrClF
MDL Number MFCD00070747
CAS 33863-76-2
Beilstein 2432912
Quantity 5g
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.