missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Bromo-3-chloro-2-methylpropane, TCI America™

$56.53 - $279.59

Chemical Identifiers

CAS 6974-77-2
Molecular Formula C4H8BrCl
Molecular Weight (g/mol) 171.462
MDL Number MFCD00000955
InChI Key ZKDOQFPDSUOLGF-UHFFFAOYSA-N
PubChem CID 95566
IUPAC Name 1-bromo-3-chloro-2-methylpropane
SMILES CC(CCl)CBr
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B185925G View Documents TCI America
B185925G
25g Each for $56.53
 
B1859500G View Documents TCI America
B1859500G
500g Each for $279.59
 
Specifications

Chemical Identifiers

6974-77-2
171.462
ZKDOQFPDSUOLGF-UHFFFAOYSA-N
1-bromo-3-chloro-2-methylpropane
C4H8BrCl
MFCD00000955
95566
CC(CCl)CBr

Specifications

154°C
25g
C4H8BrCl
ZKDOQFPDSUOLGF-UHFFFAOYSA-N
1-bromo-3-chloro-2-methylpropane
95566
Clear Liquid at 20°C
1-Bromo-3-chloro-2-methylpropane
Colorless
6974-77-2
MFCD00000955
CC(CCl)CBr
171.462
171.46
≥98.0% (GC)
Safety and Handling

Safety and Handling

TSCA : No