Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Bromo-3,5-dinitrobenzene, 98%

Chemical Identifiers

Chemical Name or Material 1-Bromo-3,5-dinitrobenzene
CAS 18242-39-2
Formula Weight 247
MDL Number MFCD00156596
UN Number UN3459
Beilstein 2053146
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6677403

  SDS
alfa aesar™
H6677403
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6677406

  SDS
alfa aesar™
H6677406
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 247
CAS 18242-39-2
MDL Number MFCD00156596
Health Hazard 1 Warning
Chemical Name or Material 1-Bromo-3,5-dinitrobenzene
Assay 98%
Color Off-white
Health Hazard 2 GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Beilstein 2053146
Physical Form Powder
Quantity 1g
DOT Information Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: NITROBROMOBENZENES, SOLID
UN Number UN3459

Applications
It is an important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.