missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Bromo-3,5-dimethoxybenzene, 97%

Manufacturer:  Alfa Aesar™ H5531214

 View more versions of this product

Catalog No. AAH5531214



Description

Description

Applications
1-Bromo-3,5-dimethoxybenzene is used as an intermediate in the synthetic preparation of pharmaceutical inhibitors via cross-coupling reactions.

Solubility
Soluble in methanol.

Notes
Reacts with strong oxidizing agents.
Specifications

Specifications

1-Bromo-3,5-dimethoxybenzene
25g
C8H9BrO2
1,3-dimethoxy-5-bromobenzene
KRWRFIMBWRVMKE-UHFFFAOYSA-N
1-bromo-3,5-dimethoxybenzene
639187
97%
62°C to 66°C
20469-65-2
MFCD06200685
Soluble in methanol.
COC1=CC(=CC(=C1)Br)OC
217.062
217.06

Chemical Identifiers

20469-65-2
217.062
KRWRFIMBWRVMKE-UHFFFAOYSA-N
639187
COC1=CC(=CC(=C1)Br)OC
C8H9BrO2
MFCD06200685
1,3-dimethoxy-5-bromobenzene
1-bromo-3,5-dimethoxybenzene
Safety and Handling

Safety and Handling

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents