Please login to your online account to display your discounted pricing

1-Bromo-3,5-difluorobenzene, 99%, ACROS Organics™

$74.93 - $74.93

Chemical Identifiers

Chemical Name or Material 1-Bromo-3, 5-difluorobenzene
CAS 461-96-1
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C6H3BrF2
Linear Formula BrC6H3F2
Formula Weight 192.99
MDL Number MFCD00010305
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC249390100

  SDS
thermo fisher scientific
249390100
Glass bottle 10mL Each for $74.93

AC249390025

  SDS
thermo fisher scientific
249390025
Glass bottle 2.5mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥98.5%
Physical Form Liquid
Formula Weight 192.99
Quantity 10mL
Color Clear colorless to light yellow
Beilstein 05, 679
CAS Max % 100.0
Chemical Name or Material 1-Bromo-3, 5-difluorobenzene
Refractive Index 1.4980 to 1.5000 (20°C, 589nm)
Assay Percent Range 99%
Molecular Formula C6H3BrF2
Linear Formula BrC6H3F2
Infrared Spectrum Authentic
MDL Number MFCD00010305
CAS 461-96-1
CAS Min % 98.5
Health Hazard 1 Warning
Packaging Glass bottle
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
May cause damage to organs through prolonged or repeated exposure.
May cause an allergic skin reaction.
Causes skin irritation.
Very toxic to aquatic life with long lasting effects.
Flammable liquid and vapor.
Health Hazard 3 GHS P Statement:
Wear eye protection/face protection.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.